Diethyl Pyrocarbonate, >97%, Ultrapure
Diethyl Pyrocarbonate, >97%, Ultrapure
Diethyl Pyrocarbonate, >97%, Ultrapure
Thermo Scientific Chemicals

Diethyl Pyrocarbonate, >97%, Ultrapure

CAS: 1609-47-8 | C6H10O5 | 162.141 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Plastic bottle
Catalog number J14710.AC
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Quantity:
25 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
160.65
Online Exclusive
178.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS1609-47-8
IUPAC Namediethyl dicarbonate
Molecular FormulaC6H10O5
InChI KeyFFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILESCCOC(=O)OC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormClear, colorless liquid
Density @20°C1.121 ? 0.005 gm/ml
Refractive Index(20?C): 1.398 ? 0.005
Assay (unspecified)≥ 97%
  • A strong, nonspecific inhibitor of ribonuclease
  • Reacts with the N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Strong, nonspecific inhibitor of ribonuclease
  • Reacts with N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer
RUO – Research Use Only