Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure
Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure
Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure
Thermo Scientific Chemicals

Dithiothreitol, >99.5%, Molecular Biology Grade, Ultrapure

Used as protective agent for SH groups | CAS: 3483-12-3 | C4H10O2S2 | 154.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number J15397.14
Price (USD)
521.65
Online Exclusive
579.00
Save 57.35 (10%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
521.65
Online Exclusive
579.00
Save 57.35 (10%)
Each
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Absorbance (280 nm, 0.1M, 1 cm): ≤ 0.08
Impurity contentOxidized DTT: ≤ 0.3%
Protease activityPasses test
Assay (unspecified)(SH): ≥ 99.5%
FormWhite crystalline powder
View more
Used as a protective agent for SH groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only