Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
J17886.1425 gPlastic bottle
J17886.031 gPlastic bottle
J17886.065 gPlastic bottle
J17886.22100 gPlastic bottle
Catalog number J17886.14
Price (USD)
583.65
Online Exclusive
648.00
Save 64.35 (10%)
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
583.65
Online Exclusive
648.00
Save 64.35 (10%)
Each
Chemical Identifiers
CAS367-93-1
IUPAC Name(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(propan-2-ylsulfanyl)oxane-3,4,5-triol
Molecular FormulaC9H18O5S
InChI KeyBPHPUYQFMNQIOC-NXRLNHOXSA-N
SMILESCC(C)S[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-33° to -29° (c=1 in water)
Appearance (Color)White to pale cream
Assay from Suppliers CofA≥99% (HPLC)
Assay from Suppliers CofADioxane: ≤ 0.02% (GC)
Water Content (Karl Fischer Titration)≤ 2.0%
View more
  • Induces the lac operon in E. coli by binding and inactivating the lac repressor.
  • Selection for lac Y mutants.
  • Selection for recombinant plasmids containing β-galactosidase fusion proteins.
  • Induces the cellular content of lactose permease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Induces lac operon in E. coli by binding and inactivating lac repressor
  • Selection for lac Y mutants
  • Selection for recombinant plasmids containing beta-galactosidase fusion proteins
  • Induces cellular content of lactose permease
RUO – Research Use Only