MOPS, ≥98.5% (Dry Basis), Ultrapure
MOPS, ≥98.5% (Dry Basis), Ultrapure
MOPS, ≥98.5% (Dry Basis), Ultrapure
Thermo Scientific Chemicals

MOPS, ≥98.5% (Dry Basis), Ultrapure

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Catalog number J19256.A1
Price (USD)
333.65
Online Exclusive
370.00
Save 36.35 (10%)
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
333.65
Online Exclusive
370.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS1132-61-2
IUPAC Name4-(3-sulfonatopropyl)morpholin-4-ium
Molecular FormulaC7H15NO4S
InChI KeyDVLFYONBTKHTER-UHFFFAOYSA-N
SMILES[O-]S(=O)(=O)CCC[NH+]1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormWhite/clear crystalline powder
Assay (unspecified)Absorbance (250 nm, 1M, 1 cm): ≤ 0.1
Impurity contentSulfate (SO₄): ≤ 0.02%
pKa(20?C): 7.20 ? 0.15
Residue on ignition≤ 0.1%
View more
RUO – Research Use Only