Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals
Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals
Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nonidet™ P40 Substitute, Ultrapure, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
J19628.K21 LPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J19628.K2
Price (USD)
-
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9002-93-1
IUPAC Name2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
InChI KeyJYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILESCC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
Synonymtriton X polyoxyethylene (11) octyl phenyl ether polyoxyethylene (9) octylphenyl ether hydrol SW poletoxol peg-12 octyl phenyl ether triton X-102 ortho-gynol octoxynol octylphenoxypolyethoxyethanol
View more
SpecificationsSpecification SheetSpecification Sheet
FormClear, slightly yellow-green viscous liquid
Optical Rotation1.06 ? 0.005
RUO – Research Use Only