D-(+)-Raffinose Pentahydrate
D-(+)-Raffinose Pentahydrate
D-(+)-Raffinose Pentahydrate
Thermo Scientific Chemicals

D-(+)-Raffinose Pentahydrate

CAS: 17629-30-0 | C18H42O21 | 594.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number J21060.22
Price (USD)
245.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
245.00
Each
Chemical Identifiers
CAS17629-30-0
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R,6R)-6-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-3,4,5-trihydroxyoxan-2-yl]methoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC18H32O16
InChI KeyMUPFEKGTMRGPLJ-ZQSKZDJDSA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO[C@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormWhite crystalline powder
Loss on Drying≤ 15.3%
Trace MetalArsenic (As): ≤ 0.5 ppm
Trace MetalHeavy Metals (Cu): ≤ 10 ppm
Optical Rotation+104.9? ? 1.3? (c=4 in H₂O & NH₃) (corrected to a theoretical pentahydrate)
View more
RUO – Research Use Only