Tricine, ≥98.5% (Dry Basis), Ultrapure
Tricine, ≥98.5% (Dry Basis), Ultrapure
Tricine, ≥98.5% (Dry Basis), Ultrapure
Thermo Scientific Chemicals

Tricine, ≥98.5% (Dry Basis), Ultrapure

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number J22561.A1
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Chemical Identifiers
CAS5704-04-1
IUPAC Name2-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}acetic acid
Molecular FormulaC6H13NO5
InChI KeySEQKRHFRPICQDD-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤ 0.5%
pKa(20?C): 8.15 ? 0.15
Trace MetalIron (Fe): ≤ 30 ppm
FormWhite/clear crystalline powder
Assay (unspecified)Absorbance (250 nm, 5%, 1 cm): ≤ 0.1
View more
RUO – Research Use Only