Tris, 99.0 to 101.0% (Dry Basis)
Tris, 99.0 to 101.0% (Dry Basis)
Tris, 99.0 to 101.0% (Dry Basis)
Thermo Scientific Chemicals

Tris, 99.0 to 101.0% (Dry Basis)

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number J22674.A7
Price (USD)
483.00
Each
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
483.00
Each
Chemical Identifiers
CAS77-86-1
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC4H11NO3
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
SMILESNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Absorbance (250 nm, 1M, 1 cm): ≤ 0.1
Residue on ignition≤ 0.1%
Loss on Drying≤ 1%
Melting Point168? - 178?C
FormWhite/clear crystalline powder
View more
RUO – Research Use Only