What is the solubility of Peroxidase, horseradish (Cat. No. J60026.03, J60026.MC)?
Peroxidase, horseradish is soluble in water or 100mM phosphate buffer.
Search
View additional product information for Peroxidase, horseradish - FAQs (J60026.MC, J60026.03)
1 product FAQs found