Methylguanidine hydrochloride, 98%
Methylguanidine hydrochloride, 98%
Methylguanidine hydrochloride, 98%
Thermo Scientific Chemicals

Methylguanidine hydrochloride, 98%

CAS: 22661-87-6 | C2H8ClN3 | 109.557 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number J60033.06
also known as J60033-06
Price (USD)
81.65
Online Exclusive
90.40
Save 8.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
81.65
Online Exclusive
90.40
Save 8.75 (10%)
Each
Specifications
CAS22661-87-6
Chemical Name or MaterialMethylguanidine hydrochloride
ColorWhite
EINECS Number245-147-3
Formula Weight109.56
View more
Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylguanidine hydrochloride can be used in the synthesis of biaryl derivatives which were evaluated as BACE1 inhibitors.

Solubility
Soluble in water (50 mg/ml - clear, colorless to light yellow solution).

Notes
Hygroscopic, store away form water/moisture. Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
RUO – Research Use Only

General References:

  1. Tadashi. Inagami; Sheldon S. York. Effect of alkyl guanidines and alkyl amines on trypsin catalysis. Biochemistry, 1968, 7(11), 4045-4052.
  2. GuÈluÈzar Yildiz; A. Tuncay DemiryuÈrek; Inci Sahin-Erdemli; and Ilker Kanzik. Comparison of antioxidant activities of aminoguanidine, methylguanidine and guanidine by luminol-enhanced chemiluminescence. British Journal of Pharmacology, 1998 124, 905-910.