Abamectin
Abamectin
Abamectin
Thermo Scientific Chemicals

Abamectin

Interacts with GABA receptors to cause chloride channel activation | CAS: 71751-41-2 | C48H72O14
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60039.03
also known as J60039-03
Price (USD)
121.00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
121.00
Each
Add to cart
Chemical Identifiers
CAS71751-41-2
IUPAC Name(1'R,2S,4'S,5S,6R,8'R,10'E,12'S,13'S,14'E,16'E,20'R,21'R,24'S)-21',24'-dihydroxy-12'-{[(2R,4S,5S,6S)-5-{[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy}-4-methoxy-6-methyloxan-2-yl]oxy}-5,11',13',22'-tetramethyl-6-(propan-2-yl)-5,6-dihydro-3',7',19'-trioxaspiro[pyran-2,6'-tetracyclo[15.6.1.1⁴,⁸.0²⁰,²⁴]pentacosane]-10',14',16',22'-tetraen-2'-one; (1'R,2S,4'S,5S,6R,8'R,10'E,12'S,13'S,14'E,16'E,20'R,21'R,24'S)-6-(butan-2-yl)-21',24'-dihydroxy-12'-{[(2R,4S,5S,6S)-5-{[(2S,4S,5S,6S)-5-hydroxy-4-methoxy-6-methyloxan-2-yl]oxy}-4-methoxy-6-methyloxan-2-yl]oxy}-5,11',13',22'-tetramethyl-5,6-dihydro-3',7',19'-trioxaspiro[pyran-2,6'-tetracyclo[15.6.1.1⁴,⁸.0²⁰,²⁴]pentacosane]-10',14',16',22'-tetraen-2'-one
Molecular FormulaC95H142O28
InChI KeyIBSREHMXUMOFBB-MVGRHBATSA-N
SMILESCO[C@H]1C[C@H](O[C@H]2[C@H](C)O[C@H](C[C@@H]2OC)O[C@H]2[C@@H](C)C=CC=C3/CO[C@@H]4[C@H](O)C(C)=C[C@@H](C(=O)O[C@H]5C[C@@H](CC=C2/C)O[C@@]2(C5)O[C@H](C(C)C)[C@@H](C)C=C2)[C@]34O)O[C@@H](C)[C@@H]1O.CCC(C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](CC=C(C)[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@@H](O)[C@H](C)O5)[C@H](C)O4)[C@@H](C)C=CC=C4/CO[C@@H]5[C@H](O)C(C)=C[C@@H](C(=O)O3)[C@]45O)O2)C=C[C@@H]1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)Avermectin B1: 75% min.
Assay (HPLC)Avermectin B1a: 70% min.
Assay (unspecified)0.2% max.
Loss on Drying2.0% max.
Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Interacts with GABA receptors to cause chloride channel activation. Also used to control a range of agronomic, fruit, vegetable and ornamental crops, and it is used by homeowners for control of fire ants and in veterinary antihelmintic.

Solubility
Soluble in DMSO

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. H. Diserens; M. Henzelin. Determination of abamectin residues in fruits and vegetables by high-performance liquid chromatography. Journal of Chromatography A.1999833(1), 13-18.
  2. Andréa Novelli; Bruna Horvath Vieira; Daniela Cordeiro; Luciana Teresa Dias Cappelini; Eny Maria Vieira; Evaldo Luiz Gaeta Espíndola. Lethal effects of abamectin on the aquatic organismsDaphnia similis,Chironomus xanthusandDanio rerio. Chemosphere.20121(86), 36-40.