N-Lauroylsarcosine sodium salt, 95%
N-Lauroylsarcosine sodium salt, 95%
N-Lauroylsarcosine sodium salt, 95%
Thermo Scientific Chemicals

N-Lauroylsarcosine sodium salt, 95%

CAS: 137-16-6 | C15H28NNaO3 | 293.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number J60040.18
also known as J60040-18
Price (USD)
295.00
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
295.00
Each
Chemical Identifiers
CAS137-16-6
IUPAC Namesodium 2-(N-methyldodecanamido)acetate
Molecular FormulaC15H28NNaO3
InChI KeyKSAVQLQVUXSOCR-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay (unspecified)≥94.0%
Melting Point43.0-48.0°C

N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein's; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Lauroylsarcosine is used for solubilization and separation of membrane proteins and glycoprotein′s; reported to inhibit hexokinase. It is useful in concentrated salt solutions used in the cell lysis step during RNA purification (helps avoid excessive foaming). It has been used to indicate paramagnetic anisotropy sign change in micelle mesophage. Inhibits bacterial flora of human saliva/gut at 0.25% as well as acting as a fungi static agent in aqueous dispersion (1%).

Solubility
Soluble in water (293 g/L).

Notes
Hygroscopic, store away form water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent and reducing agent.
RUO – Research Use Only

General References:

  1. P Gariglio; J Buss; M H Green. Sarkosyl activation of RNA polymerase activity in mitotic mouse cells. FEBS Letters, 1974, 44(3), 330-333.
  2. R J MacDonald; G H Swift; A E Przybyla; J M Chirgwin. Isolation of RNA using guanidinium salts. Methods in Enzymology, 1987 152 219-227.