Sodium borate, 0.5M buffer soln., pH 8.0
Sodium borate, 0.5M buffer soln., pH 8.0
Sodium borate, 0.5M buffer soln., pH 8.0
Thermo Scientific Chemicals

Sodium borate, 0.5M buffer soln., pH 8.0

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60068.AK
also known as J60068-AK
Price (USD)
89.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
89.00
Each
Chemical Identifiers
CAS1330-43-4
IUPAC Namedisodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Na2O7
InChI KeyNXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
pH7.85-8.15, 10% in water
It is a component of many detergents, cosmetics, and enamel glazes. It is also used to make buffer solutions in biochemistry, as a fire retardant, as an anti-fungal compound for fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a precursor for other boron compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a component of many detergents, cosmetics, and enamel glazes. It is also used to make buffer solutions in biochemistry, as a fire retardant, as an anti-fungal compound for fiberglass, as a flux in metallurgy, neutron-capture shields for radioactive sources, a texturing agent in cooking, and as a precursor for other boron compounds.

Solubility
Fully miscible with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Kate E Archibald; Larry J Minter; Gregory A Lewbart; C Scott Bailey. Collection and characterization of semen from Chilean rose tarantulas (Grammostola rosea). American Journal of Veterinary Research, 2014, 197(2), 251-257.
  2. Mitsuru Abo; Li-Ping He; Kae Sato; Akira Okubo. Determination of monosaccharides derivatized with 2-aminobenzoic Acid by capillary electrophoresis. Methods in Molecular Biology, 2013,98445-50.