Tacrine hydrochloride
Tacrine hydrochloride
Tacrine hydrochloride
Thermo Scientific Chemicals

Tacrine hydrochloride

A potent centrally acting anticholinesterase | CAS: 1684-40-8 | C13H15ClN2 | 234.73 g/mol
Have Questions?
Catalog NumberQuantity
J60070.06
also known as J60070-06
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60070.06
also known as J60070-06
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
213.65
Special offer
Online exclusive
Ends: 30-Jun-2026
251.00
Save 37.35 (15%)
Each
Chemical Identifiers
CAS1684-40-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Purity≥97.0%
FormPowder or crystals or crystalline powder
A potent centrally acting anticholinesterase for therapy of memory deficits in patients with Alzheimer's disease. THA also selectively blocks potassium channels in the central nervous system, which results in an increased release of acetylcholine and a prolongation of the action potential of the presynaptic cholinergic neurons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in aqueous buffers

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Shuai Qian; Lisi He; Marvin Mak; Yifan Han; Chun-Yu Ho; Zhong Zuo. Synthesis, biological activity, and biopharmaceutical characterization of tacrine dimers as acetylcholinesterase inhibitors. International Journal of Pharmaceutics, 2014, 477(1-2), 442-453.
  2. Jinggong Liu et. al. Synthesis and characterization of 1H-phenanthro[9,10-d]imidazole derivatives as multifunctional agents for treatment of Alzheimer's disease. Biochimica et Biophysica Acta (BBA) - General Subjects, 2006,1840(9).