Palatinose hydrate, 98+%
Palatinose hydrate, 98+%
Palatinose hydrate, 98+%
Palatinose hydrate, 98+%
Thermo Scientific Chemicals

Palatinose hydrate, 98+%

CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J60091.22
also known as J60091-22
Price (USD)
89.65
Online Exclusive
98.90
Save 9.25 (9%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
89.65
Online Exclusive
98.90
Save 9.25 (9%)
Each
Add to cart
Chemical Identifiers
CAS343336-76-5
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4S,5S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyPVXPPJIGRGXGCY-VVLSOAGWNA-N
SMILESOC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Melting Point118.0-127.0°C
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥98.0%
Water Content≤6.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only