BioAcryl-P (30%, 19:1)
BioAcryl-P (30%, 19:1)
BioAcryl-P (30%, 19:1)
Thermo Scientific Chemicals

BioAcryl-P (30%, 19:1)

Premixed solution for preparing protein PAGE gels
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J60126.K2
also known as J60126-K2
Price (USD)
213.65
Online Exclusive
237.00
Save 23.35 (10%)
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
213.65
Online Exclusive
237.00
Save 23.35 (10%)
Each
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 19:1
Concentration30% soln
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
Health Hazard 1H302+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
View more
Premixed solution for preparing protein PAGE gels

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only