5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside

Substrate for beta-galactosidase | CAS: 107021-38-5 | C14H15BrClNO6 | 408.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number J60151.03
also known as J60151-03
Price (USD)
399.65
Special offer
Online exclusive
Ends: 31-Dec-2025
470.00
Save 70.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
399.65
Special offer
Online exclusive
Ends: 31-Dec-2025
470.00
Save 70.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS107021-38-5
IUPAC Name2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC14H15BrClNO6
InChI KeyOPIFSICVWOWJMJ-UHFFFAOYNA-N
SMILESOCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Water Content (Karl Fischer Titration)≤1.0%
Optical Rotation+147° ±2° (c=1, 1:1 DMF:water)
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
View more
Substrate for beta-galactosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only