Sodium phosphate, 0.5M buffer soln., pH 7.6
Thermo Scientific Chemicals

Sodium phosphate, 0.5M buffer soln., pH 7.6

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60158.AK
also known as J60158-AK
Price (USD)
114.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
114.00
Each
Specifications
Chemical Name or MaterialSodium phosphate buffer soln.
MDL NumberMFCD00146206
Recommended StorageAmbient temperatures
TSCAYes
Concentration0.5 M
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only