Lactulose, 99%
Lactulose, 99%
Lactulose, 99%
Thermo Scientific Chemicals

Lactulose, 99%

A synthetic sugar | CAS: 4618-18-2 | C12H22O11 | 342.297 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
Catalog number J60160.14
also known as J60160-14
Price (USD)
204.65
Special offer
Online exclusive
Ends: 28-Dec-2026
241.00
Save 36.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
204.65
Special offer
Online exclusive
Ends: 28-Dec-2026
241.00
Save 36.35
Each
Chemical Identifiers
CAS4618-18-2
IUPAC Name(2S,3R,4S,5R,6R)-2-{[(2R,3S,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyJCQLYHFGKNRPGE-FCVZTGTOSA-N
SMILESOC[C@H]1O[C@](O)(CO)[C@@H](O)[C@@H]1O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤ 2%
Appearance (Color)White
Assay (HPLC)≥ 99%
FormPowder
It is not absorbed in the gut and is frequently used in the treatment of constipation and hepatic encephalopathy. actulose is also a food ingredient, better known as galactofructose, with sweet taste and offering beneficial health benefits on digestive health. Also used as a test of small intestine bacterial overgrowth (SIBO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (76.4%) at 30°C, water (81%) at 60°C, and water (>86%) at 90°C and methanol.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Tim Vanuytsel et. al. Psychological stress and corticotropin-releasing hormone increase intestinal permeability in humans by a mast cell-dependent mechanism. Gut, 2014, 63(8), 1293-1299.
  2. C P Kelly et. al. Larazotide acetate in patients with coeliac disease undergoing a gluten challenge: a randomised placebo-controlled study. Alimentary Pharmacology and Therapeutics, 2013,37,(2), 252-262.