Loratadine, 98+%
Loratadine, 98+%
Loratadine, 98+%
Thermo Scientific Chemicals

Loratadine, 98+%

A nonsedating-type histamine H1-receptor antagonist | CAS: 79794-75-5 | C22H23ClN2O2 | 382.888 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60190.03
also known as J60190-03
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
174.00
Save 26.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
147.65
Special offer
Online exclusive
Ends: 30-Jun-2026
174.00
Save 26.35 (15%)
Each
Chemical Identifiers
CAS79794-75-5
IUPAC Nameethyl 4-{13-chloro-4-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-ylidene}piperidine-1-carboxylate
Molecular FormulaC22H23ClN2O2
InChI KeyJCCNYMKQOSZNPW-UHFFFAOYSA-N
SMILESCCOC(=O)N1CCC(CC1)=C1C2=CC=C(Cl)C=C2CCC2=CC=CN=C12
View more
SpecificationsSpecification SheetSpecification Sheet
CommentHeavy metals: 20ppm max.
Assay (HPLC)98.0% min.
Loss on Drying0.5% max.
Ash20ppm max.
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Loratadine, is used as a peripheral histamine H1 receptor agonist. It is also used as an orally active antiallergic agent. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is soluble in DMSO (50 mg/ml), ethanol (77 mg/ml at 25°C), water (<1mg/mL at 25°C), chloroform, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Barnett et al. Evaluation of the CNS properties of SCH 29851, a potential non-sedating antihistamine. Agents Actions. 1984, 14,(5-6), 590-7.
  2. Kreutner et al. Antiallergic activity of loratidine, a non-sedating antihistamine. Allergy. 1987, 42,(1), 57-63.