Sodium orthovanadate, 100mM aq. soln. - FAQs
Sodium orthovanadate, 100mM aq. soln. - FAQs
View additional product information for Sodium orthovanadate, 100mM aq. soln. - FAQs (J60191.AD, J60191.AE)
1 product FAQs found
Is Sodium orthovanadate, 100 mM aq. soln. (Cat. No. J60191.xx) activated?
Sodium orthovanadate, 100mM aq. soln. (Cat. No. J60191.xx) is activated and suitable for use as a phosphatase inhibitor. Activation was achieved by boiling and adjusting the pH to 10.0.