Is Sodium orthovanadate, 100 mM aq. soln. (Cat. No. J60191.xx) activated?
Sodium orthovanadate, 100mM aq. soln. (Cat. No. J60191.xx) is activated and suitable for use as a phosphatase inhibitor. Activation was achieved by boiling and adjusting the pH to 10.0.