Dextran, MW ca 250,000
Dextran, MW ca 250,000
Dextran, MW ca 250,000
Thermo Scientific Chemicals

Dextran, MW ca 250,000

CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60200.22
also known as J60200-22
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS9004-54-0
IUPAC NameDextran
Molecular Formula(C6H9O5)n(C6H10O5)m
SMILESO[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
MDL NumberMFCD00130935
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder
Ash≤2.0%
Loss on Drying≤9.0%
Molecular Weight240,000 to 260,000
View more
Dextran, is used as a multi-use, branched glucan. Low molecular weight dextran is useful as a stabilizer, volume extender, lubricant, binding platform, and matrix component. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only