Collagen, Bovine Achilles tendon
Thermo Scientific Chemicals
Collagen, Bovine Achilles tendon
For research use
Have Questions?
Change viewbuttonViewtableView
Quantity:
1g
10g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60218.09
also known as J60218-09
Price (USD)
-
Quantity:
10g
Request bulk or custom format
Chemical Identifiers
CAS9007-34-5
SpecificationsSpecification SheetSpecification Sheet
CommentSuitable as a substrate for collagenase
Collagen, bovine achilles tendon is suitable for cell adhesion in tissue culture, as well as gel formation, platelet aggregation, and as a substrate for collagenase. It is also been used as cell culture substratum, adhesion. It is also used as an attachment factor for primary cultures of epithelioid cells.