TE buffer, pH 7.4, RNAse free
TE buffer, pH 7.4, RNAse free
TE buffer, pH 7.4, RNAse free
TE buffer, pH 7.4, RNAse free
Thermo Scientific Chemicals

TE buffer, pH 7.4, RNAse free

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
125 mL
250 mL
Catalog number J60234.EQE
also known as J60234-=E
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.30
Save 11.65 (15%)
Each
Add to cart
Quantity:
125 mL
Request bulk or custom format
Price (USD)
64.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.30
Save 11.65 (15%)
Each
Add to cart
Specifications
CAS1185-53-1
Chemical Name or MaterialTE buffer
ColorUndesignated
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
View more
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TE buffer is a commonly used buffer solution in procedures involving DNA or RNA. It is used for storing and diluting DNA and RNA. Treatment with TE buffer enhances the staining intensity of antibodies in the immunohistochemical detection of certain proteins.

Solubility
Fully miscible in water.

Notes
200mM Tris-HCl with 20mM EDTA
RUO – Research Use Only

General References:

  1. Samuel C. Brooks; Elizabeth R. Locke and Herbert D. Soule. Estrogen Receptor in a Human Cell Line (MCF-7) from Breast Carcinoma. The Journal of Biological Chemistry. 1973, 248 (5), 6251-6253.
  2. Kevin D. Burris; Thaddeus F. Molski; Cen Xu; Elaine Ryan; Katsura Tottori; Tetsuro Kikuchi; Frank D. Yocca and Perry B. Molinoff. Aripiprazole, a Novel Antipsychotic, Is a High-Affinity Partial Agonist at Human Dopamine D2 Receptors. JPET. 2002, 302 (1), 381-389.