L-Alanine, Cell Culture Reagent
L-Alanine, Cell Culture Reagent
L-Alanine, Cell Culture Reagent
L-Alanine, Cell Culture Reagent
Thermo Scientific Chemicals

L-Alanine, Cell Culture Reagent

CAS: 56-41-7 | C3H7NO2 | 89.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Catalog number J60279.18
also known as J60279-18
Price (USD)
70.50
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
70.50
Each
Add to cart
Chemical Identifiers
CAS56-41-7
IUPAC Name(2S)-2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-REOHCLBHSA-N
SMILESC[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤0.5%
Appearance (Color)White
Assay from Suppliers CofA≥98.0%
FormPowder or crystals or crystalline powder
Residue on ignition≤0.2%
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a key player in the Glucose-Alanine cycle, which enables the removal of pyruvate and glutamate from muscle to the liver. The Glucose-Alanine cycle aides to conserve ATP in muscle for muscle contraction, while the energy burden of gluconeogenesis is imposed upon the liver.

Solubility
Soluble in water 166.5 g/L..

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Aubérie Parent; Xavier Elduque; David Cornu; Laura Belot; Jean-Pierre Le Caer; Anna Grandas; Michel B Toledano; Benoit D'Autréaux. Mammalian frataxin directly enhances sulfur transfer of NFS1 persulfide to both ISCU and free thiols. Nature Communications. 2015, 6 5686.
  2. Heidi Turkia; Heli Sirén, Merja Penttilä; Juha-Pekka Pitkänen. Capillary electrophoresis with laser-induced fluorescence detection for studying amino acid uptake by yeast during beer fermentation. Talanta. 2015, 131, 366-371.