This product is animal origin and can only be shipped within the United States
Casein Blocking Buffer, 3% in PBS
Thermo Scientific Chemicals

Casein Blocking Buffer, 3% in PBS

For Western blots and ELISA
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60289.AK
also known as J60289-AK
Price (USD)
-
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBlocking Buffer
Recommended StorageStore at -20°C
Concentration3% in PBS
For Use With (Application)Western Blot
Physical FormViscous Liquid
View more
The phosphate- and Tris-buffered saline formulations are used for blocking excess binding sites on membranes and microplates in antibody-based detection methods in ELISA, Western blotting, immunohistochemistry and other immunochemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only