Tyrphostin A23, 99%, Thermo Scientific Chemicals
Tyrphostin A23, 99%, Thermo Scientific Chemicals
Tyrphostin A23, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tyrphostin A23, 99%, Thermo Scientific Chemicals

Potent inhibitor of EGF receptor kinase activity
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
Catalog number J60308.MA
also known as J60308-MA
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS118409-57-7
IUPAC Name2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
Molecular FormulaC10H6N2O2
InChI KeyVTJXFTPMFYAJJU-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C(C#N)C#N)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.5%
Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrphostin inhibit protein tyrosine kinase and block the EGF-dependent autophosphorylation of the receptor. The inhibitors can bind to the EGF receptor kinase domain with different affinities. It also inhibits PDGFR kinase. Tyrphostin A23 has been used to inhibit agonist-induced GP IIb- IIIa activation in permeabilized platelets and in other studies to block calcium channel currents in vascular smooth muscle cells. . Tyrphostin A23 can also inhibit the internalization of (125)I-transferrin in Heb7a cells.

Solubility
Soluble in DMSO, ethanol, or DMF

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, acids, heat.
RUO – Research Use Only

General References:

  1. S Dunzendorfer; P Schratzberger; N Reinisch, C M Kähler; C J Wiedermann. Secretoneurin, a novel neuropeptide, is a potent chemoattractant for human eosinophils. Blood.1998, 91 (5), 1527-1532.
  2. A Gazit; P Yaish; C Gilon; A Levitzki. Tyrphostins I: synthesis and biological activity of protein tyrosine kinase inhibitors. Journal of Medicinal Chemistry. 1989, 32, (10), 2344-2352.