Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7
Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7
Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7
Thermo Scientific Chemicals

Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7

Useful for purifying RNA | CAS: 136112-00-0 | C12H19Cl3O2 | 301.632 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
400 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60331.AE
also known as J60331-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS136112-00-0
IUPAC Name3-methylbutan-1-ol; phenol; trichloromethane
Molecular FormulaC12H19Cl3O2
InChI KeyZYWFEOZQIUMEGL-UHFFFAOYSA-N
SMILESClC(Cl)Cl.CC(C)CCO.OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
UV/VisOD 330nm ≤0.2
pH6.5-6.9 at 25°C
Appearance (Color)Colorless
FormLiquid (2-layer)
Assay (unspecified)≥99.0%
View more
Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Note: Buffer is sold separately; see 327345000.

RUO – Research Use Only