Formaldehyde, 4% in PBS
Formaldehyde, 4% in PBS
Formaldehyde, 4% in PBS
Formaldehyde, 4% in PBS
Thermo Scientific Chemicals

Formaldehyde, 4% in PBS

Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60401.AK
also known as J60401-AK
Price (USD)
75.70
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)
75.70
Each
Add to cart
Chemical Identifiers
CAS50-00-0
IUPAC Nameformaldehyde
Molecular FormulaCH2O
InChI KeyWSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILESC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
CommentFormaldehyde solution is made by dissolving it in 1X PBS, pH 7.4, followed by submicron filtration
pH7.40 ± 0.20
Formaldehyde, used as an embaliming agent due to its ability to crosslink primary amino groups in proteins through a methylene linkage, Formaldehyde is also used in numerous industrial applications and organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only