Carboplatin
Carboplatin
Carboplatin
Thermo Scientific Chemicals

Carboplatin

CAS: 41575-94-4 | C6H12N2O4Pt | 371.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J60433.03
also known as J60433-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS41575-94-4
IUPAC Name5,9-dioxo-6,8-dioxa-7-platinaspiro[3.5]nonane-7,7-bis(ylium) diamine
Molecular FormulaC6H12N2O4Pt
InChI KeyBHKICZDKIIDMNR-UHFFFAOYSA-L
SMILESN.N.O=C1O[Pt++]OC(=O)C11CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (HPLC)98.0% min.
FormPowder
Carboplatin is primarily used as an antitumor agent.Carboplatin is widely utilized as a chemotherapy drug to treat various types of cancer. It belongs to the group of platinum-based antineoplastic agents to interfere in DNA (deoxyribonucleic acid) repair. It is applied in combination other medications and used to inhibit cancer cell growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Jakacki, R. I.; Burger, P. C.; Kocak, M.; Boyett, J. M.; Goldwein, J.; Mehta, M.; Packer, R. J.; Tarbell, N. J.; Pollack, I. F. Outcome and Prognostic Factors for Children With Supratentorial PrimitiveNeuroectodermal Tumors Treated With Carboplatin During Radiotherapy:A Report from the Children’s Oncology Group. Pediatr Blood Cancer 2015, 62 (5), 776-783.
  2. Boal, A. K.; Rosenzweig, A. C. Response from Boal and Rosenzweig to Crystallography and chemistry should always go together: a cautionary tale of protein complexes with cisplatin and carboplatin. Acta Crystallogr., Sect. D: Biol. Crystallogr. 2015, 71, 1984-1986.