alpha-D-(+)-Melibiose, Thermo Scientific Chemicals
alpha-D-(+)-Melibiose, Thermo Scientific Chemicals
alpha-D-(+)-Melibiose, Thermo Scientific Chemicals
Thermo Scientific Chemicals

alpha-D-(+)-Melibiose, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
J60439.06
also known as J60439-06
5 g
Catalog number J60439.06
also known as J60439-06
Price (USD)
210.65
Online Exclusive
234.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
210.65
Online Exclusive
234.00
Save 23.35 (10%)
Each
Specifications
CAS585-99-9
Chemical Name or Materialalpha-D-(+)-Melibiose
ColorWhite
EINECS Number209-568-6
Formula Weight342.3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only