Phosphate-buffered saline (PBS, 10X), no potassium
Thermo Scientific Chemicals

Phosphate-buffered saline (PBS, 10X), no potassium

Helps to maintain a constant pH
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J60465.AP
also known as J60465-AP
Price (USD)
92.40
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
92.40
Each
Specifications
Chemical Name or MaterialPhosphate Buffered Saline (PBS)
Recommended StorageAmbient temperatures
Concentration10 X
Physical FormLiquid
Quantity500 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only