Ginkgolide B
Ginkgolide B
Ginkgolide B
Thermo Scientific Chemicals

Ginkgolide B

Platelet-Activating Factor (PAF) antagonist | CAS: 15291-77-7 | C20H24O10 | 424.402 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
Catalog number J60646.MF
also known as J60646-MF
Price (USD)
326.65
Special offer
Online exclusive
Ends: 30-Jun-2026
384.00
Save 57.35 (15%)
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
326.65
Special offer
Online exclusive
Ends: 30-Jun-2026
384.00
Save 57.35 (15%)
Each
Chemical Identifiers
CAS15291-77-7
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% min.
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ginkgolide B is used as a potent PAF-R antagonist. Ginkgolide B speeds up PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Additionally, this compound has been reported to inhibit neutrophil degranulation in vitro. Inhibits superoxide production in vitro and inhibits bronchoconstriction.

Solubility
Soluble in DMSO. Slightly soluble in water and ethanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Desquand et al. Interference of BN 52021 (ginkgolide B) with the bronchopulmonary effects of PAF-acether in the guinea-pig. Eur.J.Pharmacol. 1986, 127 ,(1-2), 83-95.
  2. Foldes-Filep et al. Inhibition by BN 52021 (ginkgolide B) of the binding of [3H]-platelet-activating factor to human neutrophil granulocytes. Biochem.Biophys.Res.Commun. 1987, 148,(3), 1412-7.