PIPES, 1.0M buffer soln., pH 6.5
PIPES, 1.0M buffer soln., pH 6.5
PIPES, 1.0M buffer soln., pH 6.5
Thermo Scientific Chemicals

PIPES, 1.0M buffer soln., pH 6.5

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60659.AK
also known as J60659-AK
Price (USD)
163.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
163.00
Each
Specifications
CAS5625-37-6
ChEBICHEBI:44933
Chemical Name or MaterialPIPES buffer soln.
ColorColorless
IUPAC Name2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
View more

PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES was used in the preparation of a variety of marine and mammalian tissues for electron microscopic study. Lipid loss and artifacts are minimized when PIPES was used to buffer the glutaraldehyde fixative. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer. Alkaline phosphatase retention activity was 60% greater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
PIPES has pKa near the physiological pH which makes it useful in cell culture work. PIPES was used in the preparation of a variety of marine and mammalian tissues for electron microscopic study. Lipid loss and artifacts are minimized when PIPES was used to buffer the glutaraldehyde fixative. Fixation of fungal zoospores for fluorescence microscopy and electron microscopy was optimal with a combination of glutaraldehyde and formaldehyde in PIPES buffer. Alkaline phosphatase retention activity was 60% greater.

Solubility
Fully misicible in water. Soluble in water (0.05 M), giving a clear, colorless solution (the resulting solution has a pH of approximately 4.5-4.7). 10 g will dissolve in 40 mL of 0.1 N NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, reducing agents, heat.
RUO – Research Use Only

General References:

  1. P. S. Baur,; T. R. Stacey.The use of PIPES buffer in the fixation of mammalian and marine tissues for electron microscopy. Journal of Microscopy. 1977 , 109 (3), 315-327.
  2. K. Yamamoto,; K. Ogawa. Effects of NaOH-PIPES buffer used in aldehyde fixative on alkaline phosphatase activity in rat hepatocytes. Histochemistry. 1983, 77 (3), 339-351.