Hygromycin B
Hygromycin B
Hygromycin B
Thermo Scientific Chemicals

Hygromycin B

Hygromycin B, CAS # 31282-04-9, is an aminoglycoside compound showing antibiotic activity that kills bacteria through the inhibition of protein synthesis. It also presents anthelmintic activity. | CAS: 31282-04-9 | C20H37N3O13 | 527.524 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60681.MD
also known as J60681-MD
Price (USD)
138.65
Special offer
Online exclusive
Ends: 30-Jun-2026
163.00
Save 24.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
138.65
Special offer
Online exclusive
Ends: 30-Jun-2026
163.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS31282-04-9
IUPAC Name(3'R,3aS,4S,4'R,5'R,6R,6'R,7S,7aS)-4-{[(1R,2S,3R,5S,6R)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy}-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)-tetrahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
Molecular FormulaC20H37N3O13
InChI KeyGRRNUXAQVGOGFE-KPBUCVLVSA-N
SMILESCN[C@H]1C[C@@H](N)[C@H](O)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H]3OC4(O[C@H]23)O[C@H]([C@@H](N)CO)[C@H](O)[C@@H](O)[C@H]4O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to plale brown
FormPowder
Assay (HPLC)≥ 90.0%
Water Content (Karl Fischer Titration)15.0%
Potency≥ 900 units/ mg
View more
An aminoglycoside antibiotic that kills bacteria by inhibiting protein synthesisHygromycin B is used for the selection and maintenance of prokaryotic and eukaryotic cells that contains the hygromycin resistance gene. This gene is frequently used as a selectable marker in research on plants. It stabilizes the transfer ribonucleic acid (tRNA)-ribosomal acceptor site, thereby inhibiting translocation. In addition to this, it serves as an aminoglycosidic antibiotic which inhibits protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hygromycin is an aminoglycosidic compound that shows antibiotic activity. It is produced by Streptomyces hygroscopicus
  • Hygromycin acts against both prokaryotic and eukaryotic cells, having the ability to inhibit protein synthesis. It also stabilizes the transfer ribonucleic acid (tRNA)-ribosomal acceptor site, thereby inhibiting translocation. This compound prevents ribosome translocation during translation elongation by binding the 30S rRNA proximal to the ribosomal E, P, and A sites

Application

  • In clinical and plant research, this compound can be used as a marker to select and maintain prokaryotic and eukaryotic cells containing the hygromycin resistance gene
  • Hygromycin also presents an anthelmintic activity against nematode parasites of chickens
RUO – Research Use Only

General References:

  1. Chen, C. F.; Chan, K. G.; Tan, B. C.; Khalid, N. Enhancement of Agrobacterium-mediated transformation efficiency of model plant using quorum sensing molecule, N-3-oxo-octanoyl-l-homoserine-lactone. Plant Cell Tissue Organ Cult. 2015, 121 (2), 481-487.
  2. Park, Y.; Hosomichi, J.; Ge, C.; Xu, J.; Franceschi, R.; Kapila, S. Immortalization and characterization of mouse temporomandibular joint disc cell clones with capacity for multi-lineage differentiation. Osteoarthr. Cartil. 2015, 23 (9), 1532-1542.