alpha-Cyclodextrin, 97+%
alpha-Cyclodextrin, 97+%
alpha-Cyclodextrin, 97+%
alpha-Cyclodextrin, 97+%
Thermo Scientific Chemicals

alpha-Cyclodextrin, 97+%

Useful for selective precipitation of enantiomeric, positional or structural isomers | CAS: 10016-20-3 | C36H60O30 | 972.846 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number J60687.22
also known as J60687-22
Price (USD)
460.65
Special offer
Online exclusive
Ends: 30-Jun-2026
542.00
Save 81.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
460.65
Special offer
Online exclusive
Ends: 30-Jun-2026
542.00
Save 81.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS10016-20-3
IUPAC Name5,10,15,20,25,30-hexakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29-dodecaoxaheptacyclo[26.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶]dotetracontane-31,32,33,34,35,36,37,38,39,40,41,42-dodecol
Molecular FormulaC36H60O30
InChI KeyHFHDHCJBZVLPGP-UHFFFAOYNA-N
SMILESOCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC1C(O)C2O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition≤ 0.1 %
Assay (Titration)≥ 97.0 %
Impurity contentOrganic volatile impurities: ≤ 20 ppm
Appearance (Color)White to pale cream
Water Content (Karl Fischer Titration)≤ 10 %
View more
Useful for selective precipitation of enantiomeric, positional or structural isomersα-Cyclodextrin is used as a fiber ingredient, an odor or flavor masking agent. It is also useful for emulsification applications. It is also used as whipping fiber and emulsifying fiber. It finds application in medical, healthcare and food and beverage applications. It is also used to lower blood low-density lipoprotein cholesterol levels and lower blood triglyceride levels. It plays an essential role in fat free or fat containing dessert compositions and also employed for the reduction or the replacement of egg white in confectionary and bakery applications. Further, it acts as a supramolecular carrier, complexing agent and controlled drug release. In addition to this, it is used to increase the insulin and leptin sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 1%(w/v)

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Guevara, M. A.; Bauer, L. L.; Garleb, K. A.; Fahey, G. C.; de Godoy, M. R. C. Serum lipid profiles, total tract nutrient digestibility, and gastrointestinal tolerance by dogs of alpha-cyclodextrin. J. Anim. Sci. 2015, 93 (5), 2201-2207.
  2. Ma, P.; Liang, F.; Wang, D.; Yang, Q.; Cao, B.; Song, D.; Gao, D.; Wang, X. Selective determination of o-phenylenediamine by surface-enhanced Raman spectroscopy using silver nanoparticles decorated with alpha-cyclodextrin. Microchim. Acta 2015, 182 (1), 167-174.