TE buffer, (20X), pH 7.4, autoclaved
TE buffer, (20X), pH 7.4, autoclaved
TE buffer, (20X), pH 7.4, autoclaved
Thermo Scientific Chemicals

TE buffer, (20X), pH 7.4, autoclaved

CAS: 1185-53-1 | C4H12ClNO3 | 157.594 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J60738.AK
also known as J60738-AK
Price (USD)
73.60
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
73.60
Each
Specifications
CAS1185-53-1
Chemical Name or MaterialTE buffer
EINECS Number231-791-2
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
InChI KeyQKNYBSVHEMOAJP-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only