Irinotecan hydrochloride
Irinotecan hydrochloride
Irinotecan hydrochloride
Thermo Scientific Chemicals

Irinotecan hydrochloride

Topoisomerase I inhibitor | CAS: 100286-90-6 | C33H39ClN4O6 | 623.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60743.MD
also known as J60743-MD
Price (USD)
752.65
Online exclusive
836.00
Save 83.35
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
752.65
Online exclusive
836.00
Save 83.35
Each
Chemical Identifiers
CAS100286-90-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.0% min.
Topoisomerase I inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only