ACES Buffer  (0.5 M, pH 7.0)
ACES Buffer  (0.5 M, pH 7.0)
ACES Buffer  (0.5 M, pH 7.0)
Thermo Scientific Chemicals

ACES Buffer (0.5 M, pH 7.0)

CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J60755.AE
also known as J60755-AE
Price (USD)
68.80
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
68.80
Each
Specifications
CAS7365-82-4
ChEBICHEBI:39062
Chemical Name or MaterialACES buffer soln.
ColorColorless
IUPAC Name2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only