CGS 21680 hydrochloride, 99%
CGS 21680 hydrochloride, 99%
Thermo Scientific Chemicals

CGS 21680 hydrochloride, 99%

An A2A Adenosine receptor agonist | CAS: 120225-54-9 | C23H30ClN7O6 | 535.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mg
10 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60793.MA
also known as J60793-MA
Price (USD)
407.65
Online Exclusive
453.00
Save 45.35 (10%)
Each
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (USD)
407.65
Online Exclusive
453.00
Save 45.35 (10%)
Each
Add to cart
Specifications
CAS120225-54-9
Chemical Name or MaterialCGS 21680 hydrochloride
ColorWhite
Formula Weight535.98
IUPAC Name3-[4-[2-[[6-amino-9-[(2S,4S,5R)-5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl]amino]ethyl]phenyl]propanoic acid;hydrochloride
View more
CGS 21680 hydrochloride is currently used in research in studying neuronal transmission, also in respiration. CGS 21680 is commonly used to study the actions of the adenosine A2A receptor in cells and tissues. CGS 21680 HCl is an adenosine A2 receptor agonist with IC50 of 22 nM, exhibits 140-fold over A1 receptor. CGS 21680C effectively increases coronary flow with an ED25 value of 1.8 nM. In hippocampal slices, CGS 21680 apis weak agonist on pre- and postsynaptic measures of electrophysiologicaJ activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CGS 21680 hydrochloride is currently used in research in studying neuronal transmission, also in respiration. CGS 21680 is commonly used to study the actions of the adenosine A2A receptor in cells and tissues. CGS 21680 HCl is an adenosine A2 receptor agonist with IC50 of 22 nM, exhibits 140-fold over A1 receptor. CGS 21680C effectively increases coronary flow with an ED25 value of 1.8 nM. In hippocampal slices, CGS 21680 apis weak agonist on pre- and postsynaptic measures of electrophysiologicaJ activity.

Solubility
Soluble up to 3.4 mg/mL in DMSO and 20 mg/mL in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cheng, M.K.,; Doumad, A.B., Jiang, H., et al. Epoxyeicosatrienoic acids mediate adenosine-induced vasodilation in rat prelomerular microvessels (PGMV) via A2A receptors. Br J Pharmacol . 2004, 141 441-448 .
  2. Phillis,J.W.. The selective adenosine A2 receptor agonist, CGS 21680, is a potent depressant of cerebral cortical neuronal activity.. Brain Res.. 1990 , 509 328-330.