Potassium phosphate, 0.2M buffer soln., pH 8.0
Potassium phosphate, 0.2M buffer soln., pH 8.0
Potassium phosphate, 0.2M buffer soln., pH 8.0
Thermo Scientific Chemicals

Potassium phosphate, 0.2M buffer soln., pH 8.0

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J60851.K2
also known as J60851-K2
Price (USD)
156.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
156.00
Each
Specifications
CAS7778-77-0
Chemical Name or MaterialPotassium phosphate
IUPAC Namepotassium dihydrogen phosphate
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
MDL NumberMFCD00011401 MFCD00147253
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only