Dantrolene sodium salt
Dantrolene sodium salt
Dantrolene sodium salt
Thermo Scientific Chemicals

Dantrolene sodium salt

Inhibits intracellular calcium release from the sarcoplasmic reticulum | CAS: 14663-23-1 | C14H12N4NaO5 | 339.263 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60887.MC
also known as J60887-MC
Price (USD)
85.10
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
85.10
Each
Specifications
CAS14663-23-1
Chemical Name or MaterialDantrolene sodium salt
ColorOrange to Yellow
EINECS Number238-706-8
Formula Weight336.23
View more
Inhibits intracellular calcium release from the sarcoplasmic reticulum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only