Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals
Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals
Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
125 mL
250 mL
Catalog number J60899.AK
also known as J60899-AK
Price (USD)
111.65
Special offer
Online exclusive
Ends: 30-Jun-2026
131.00
Save 19.35 (15%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
111.65
Special offer
Online exclusive
Ends: 30-Jun-2026
131.00
Save 19.35 (15%)
Each
Specifications
CAS127-09-3
ChEBICHEBI:32954
Chemical Name or MaterialSodium acetate
Concentration3M aq. soln.
Formula Weight82.03
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only