Search
| Catalog Number | Quantity |
|---|---|
| J60915.03 also known as J60915-03 | 1 g |
Colistin sulfate A Polymyxin potent antibiotic and apoptosis inducer. This compound induces apoptosis through interaction with the cytoplasmic membrane. Colistin is a key microbiological component in Colistin Oxolinic Acid Blood Agar utilized in the cultivation of Aminobacter aminovorans, Bacillus species, Hyphomicrobium species and Methylobacterium species. It is also a critical component is VCN Inhibitor & VCNT Inhibitor growth media used in the isolation of Neisseria species.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Applications
General References: