Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)
Thermo Scientific Chemicals

Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
50 mL
Catalog number J60933.AC
also known as J60933-AC
Price (USD)
75.40
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
75.40
Each
Specifications
Chemical Name or MaterialAgarose gel loading dye
Concentration6X
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
View more
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

Solubility
Fully misicible in water.

Notes
No Xylene cyanol FF. Contains 0.25% bromophenol blue, 30% glycerol.
RUO – Research Use Only