Ampicillin
Ampicillin
Ampicillin
Thermo Scientific Chemicals

Ampicillin

Ampicillin, CAS # 69-53-4, is structurally related to penicillin and is considered to be a semi-synthetic antibiotic with antibacterial activity. | CAS: 69-53-4 | C16H19N3O4S | 349.405 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J60977.06
also known as J60977-06
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
129.65
Online Exclusive
144.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS69-53-4
IUPAC Name(2S,5R,6R)-6-[(2R)-2-amino-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular FormulaC16H19N3O4S
InChI KeyAVKUERGKIZMTKX-NJBDSQKTSA-N
SMILESCC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)C3=CC=CC=C3)C(=O)N2[C@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)White to off-white
Water Content (Karl Fischer Titration)≤ 3.0%
Sulfated Ash≤ 0.3%
Assay from Suppliers CofA≥ 98.0% (on dry basis)
View more
Ampicillin is used as asemi-synthetic antibiotic which has antibacterial activity structurally related to penicillin. It is widely used selection reagent for transformed cells expressing β-lactamase. It is also used to prevent and treat a number of bacterial infections. It may also be used to prevent group B streptococcal infection in newborns.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ampicillin is a broad-spectrum, semi-synthetic, β-lactam penicillin with bactericidal activity. It acts against several gram-positive and -negative infections
  • This compound binds to and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall. This inactivation interferes with the cross-linkage of peptidoglycan chains essential for bacterial cell wall strength and rigidity. Consequently, there is an interruption in the bacterial cell wall synthesis which results in the weakening of the bacterial cell wall causing cell lysis

Application

  • In laboratory experiments, it is commonly used as a selector reagent for transformed cells expressing β-lactamase. The ampicillin-resistant gene (ampR) catalyzes the hydrolysis of the β-lactam ring of ampicillin and naturally detoxifies the drug
  • Ampicillin is commonly used as a selection marker for E. coli and other bacteria during plasmid DNA isolation, protein expression, and gene cloning
RUO – Research Use Only

General References:

  1. Zhao Z., Baldo BA., O'Brien RM., Plomley RF. Reaction with, and fine structural recognition of polyamines by human IgE antibodies. Molecular immunology . 2000, 37,(5), 233-240.
  2. Harmoinen J., Vaali K., Koski P., Syrjänen K., Laitinen O., Lindevall K., Westermarck E. Enzymic degradation of a beta-lactam antibiotic, ampicillin, in the gut: a novel treatment modality. The Journal of antimicrobial chemotherapy . 2003, 51,(2), 361-365.

Frequently asked questions (FAQs)

How can I prepare LB-ampicillin using Ampicillin (Cat. No. J60977)?

We recommend the following the instructions to prepare LB-ampicillin:

  1. Dissolve the following in 500 mL dH2O:
    • 5g tryptone
    • 2.5g yeast extract
    • 5.0g NaCl
    • 7.5g agar
    • OR
    • 20g pre-mixed LB agar powder
  2. Boil the solution on a hot plate while stirring for 1-2 minutes.
  3. Autoclave the solution for 20 minutes and let cool to 50-60°C.
  4. Add 25 mg ampicillin and swirl or stir on a hot plate to mix.
  5. Pour approximately 10 mL of molten LB agar into each plate.
  6. Allow plates to solidify for approximately 20 minutes.

Can Ampicillin (Cat. No. J60977), be used for administration to or consumption by humans or animals?

Ampicillin, (Cat. No. J60977), is an RUO (Research Use Only) chemical. It is not intended for animal or human diagnostic or therapeutic use.