TRIS-glycine-SDS running buffer (10X), pH 8.5 - FAQs
TRIS-glycine-SDS running buffer (10X), pH 8.5 - FAQs
View additional product information for TRIS-glycine-SDS running buffer (10X), pH 8.5 - FAQs (J61006.K7, J61006.K3, J61006.K2)
1 product FAQs found
What are the concentrations of Tris base, glycine and SDS in the TRIS-glycine-SDS running buffer (10X), pH 8.3?
The TRIS-glycine-SDS running buffer (10X), pH 8.3 contains 0.25 M Tris base, 1.92 M glycine, and 1% SDS.