Pectin Citrus
Thermo Scientific Chemicals

Pectin Citrus

CAS: 9000-69-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number J61021.36
also known as J61021-36
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS9000-69-5
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (unspecified)Arsenic: 3ppm max.
Assay (unspecified)Lead: 5ppm max.
Assay (unspecified)Methoxy groups: 6.7% min.
Loss on Drying10.0% max.
View more
Pectin Citrus is promoted in dietary supplement form as an alternative cancer treatment. It can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(RUO – Research Use Only.)  Promoted in dietary supplement form as an alternative cancer treatment, Pectin Citrus can also been used to treat prostate cancer and melanoma. It can also be used as a gelling agent, filler, and stabilizer. It Supports celluar health, promotes immune function, supports cardiovascular health, supports healthy function of multiple organs, Safely chelates heavy metals and toxins.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Jesper Harholt et. Al. Biosynthesis of pectin. Plant Physiology. 2010, 153,(2), 384-395.
  2. Robert Palin and Anja Geitmann. The role of pectin in plant morphogenesis. Biosystems. 2012, 109,(3), 397-402.