Phosphatase Inhibitor Cocktail II
Thermo Scientific Chemicals

Phosphatase Inhibitor Cocktail II

 
Have Questions?
Catalog NumberQuantity
J61022.AA5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61022.AA
Price (USD)
-
Quantity:
5 mL
Request bulk or custom format
Specifications
ColorColorless
Composition100mM sodium fluoride, 50mM sodium orthovanadate, 50mM sodium pyrophosphate, and 100mM b-glycerophosphate
Concentration100X
DescriptionInhibit broad spectrum of serine/theorine and tyrosine phosphatases
Quantity5 mL
View more
The phosphatase inhibitor cocktail improves the yields of intact proteins by adding inhibitors to enzymes that modify proteins present in cell extracts. This product has been optimized and tested for tyrosine protein phosphatases, acid and alkaline phosphatases. It has been tested on cell extracts from animal tissues including bovine liver and human placenta, and cytosolic extract of rabbit muscle. It is a featured product for Kinase Phosphatase Biology research.An inhibitor of L-isozymes of alkaline phosphatase and ser/thr protein phosphatases. It is used with tissue and cell extracts, including extracts containing detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C